Learn More
Chemglass Life Sciences Condenser, Stainless Steel, 165 mm OAL, M18 x 1.5 thread for 1-5 L bioreactors
SDP
Catalog No. 502733324
Encompass
Jacketed stainless steel condenser designed for 1-5 L bioreactors. It features a 165 mm overall length, M18 x 1.5 thread connection, electropolished 316L construction, and multiple 1/4 in hose connections for jacket and gas outlet. An internal baffle improves condensate flow, and the 12 mm stem fits adjustable probe adapters.
  • Constructed from electropolished 316L stainless steel.
  • Overall length 165 mm.
  • M18 x 1.5 threaded connection.
  • Two 1/4 in hose connections on jacketed body and 1/4 in hose connection on 90° gas outlet tube.
  • Internal stainless steel baffle.
  • 12 mm O.D. stem compatible with adjustable probe adapters (sold separately).
  • Intended for use with 1-5 L bioreactors.
Catalog No. 50-273-3324 Supplier Chemglass Life Sciences Supplier No. CLS138027
Jacketed stainless steel condenser designed for 1-5 L bioreactors. It features a 165 mm overall length, M18 x 1.5 thread connection, electropolished 316L construction, and multiple 1/4 in hose connections for jacket and gas outlet. An internal baffle improves condensate flow, and the 12 mm stem fits adjustable probe adapters.
  • Constructed from electropolished 316L stainless steel.
  • Overall length 165 mm.
  • M18 x 1.5 threaded connection.
  • Two 1/4 in hose connections on jacketed body and 1/4 in hose connection on 90° gas outlet tube.
  • Internal stainless steel baffle.
  • 12 mm O.D. stem compatible with adjustable probe adapters (sold separately).
  • Intended for use with 1-5 L bioreactors.