Learn More
Chemglass Life Sciences Condenser, Stainless Steel, M18 x 1.5 Thread, 230 mm OAL, for 7 L Bioreactors
SDP
Catalog No. 502733314
Encompass
Chemglass stainless steel condenser designed for 7 L bioreactor vessels. Constructed from 316L stainless steel with an electropolished surface finish, the condenser reduces moisture and material loss from exhaust during fermentation. It features a 12 mm stem for adapter mounting and multiple hose connections for process control.
  • Stainless steel 316L construction with electropolished surface finish.
  • 230 mm overall length (OAL).
  • 12 mm outer diameter stem for probe adapter connection.
  • M18 x 1.5 connection when used with an adjustable probe holder or adapter.
  • Two 1/4 in hose connections and 90° gas outlet with 1/4 in hose connection.
  • Includes jacketed condenser body, internal stainless steel baffle, outlet tube, and threaded compression nut.
  • Designed to minimize evaporation and reduce moisture in bioreactor exhaust during fermentation.
Catalog No. 50-273-3314 Supplier Chemglass Life Sciences Supplier No. CLS138309
Add to Cart
Add to Cart
Chemglass stainless steel condenser designed for 7 L bioreactor vessels. Constructed from 316L stainless steel with an electropolished surface finish, the condenser reduces moisture and material loss from exhaust during fermentation. It features a 12 mm stem for adapter mounting and multiple hose connections for process control.
  • Stainless steel 316L construction with electropolished surface finish.
  • 230 mm overall length (OAL).
  • 12 mm outer diameter stem for probe adapter connection.
  • M18 x 1.5 connection when used with an adjustable probe holder or adapter.
  • Two 1/4 in hose connections and 90° gas outlet with 1/4 in hose connection.
  • Includes jacketed condenser body, internal stainless steel baffle, outlet tube, and threaded compression nut.
  • Designed to minimize evaporation and reduce moisture in bioreactor exhaust during fermentation.