Learn More
Heidolph™ Condenser T Auto for Laborota 20, Hei-VAP Industrial Rotary Evaporator
SDP
Catalog No. 13900923 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Laborota 20, Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13-900-923 Laborota 20, Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13-900-923 Supplier Heidolph™ Supplier No. 036247510
Add to Cart
Add to Cart
Request a Quote
Condenser T auto allows distillate to re-condense. Identical to condenser T auto R but features a condenser that is 320 mm shorter.
Included with Laborota 20, Hei-VAP Industrial Rotary Evaporator Glassware set Shorty R and Shorty RC
Specifications
Type Glassware
For Use With (Equipment) Laborota 20, Hei-VAP Industrial Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov