Learn More
Ace Glass, Inc. Condenser, triple coil, poly-coated, model R220, glassware setsd&d2, part 27308
SDP
Catalog No. 50978235
Encompass
Borosilicate glass replacement poly-coated triple coil condenser for use in rotary evaporator Model R220 glassware sets D & D2; part 27308. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.
Catalog No. 50-978-235 Supplier Ace Glass, Inc. Supplier No. 396510
Add to Cart
Add to Cart
Request a Quote
Borosilicate glass replacement poly-coated triple coil condenser for use in rotary evaporator Model R220 glassware sets D & D2; part 27308. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.