Learn More
Ace Glass, Inc. Condenser, triple coil, poly-coated, model R220, glassware setsr&d2, part 41159
SDP
Catalog No. 50971485
Encompass
Borosilicate glass replacement poly-coated triple coil condenser for use in rotary evaporator Model R220 glassware sets R & D2; part 41159. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.
Catalog No. 50-971-485 Supplier Ace Glass, Inc. Supplier No. 396505
Borosilicate glass replacement poly-coated triple coil condenser for use in rotary evaporator Model R220 glassware sets R & D2; part 41159. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.