Learn More
Ace Glass, Inc. Condenser, triple coil, models R220EX&SE, glassware sets D, D2, part 41333
SDP
Catalog No. 50971487
Encompass
Borosilicate glass replacement triple coil condenser for use in rotary evaporator Model R220EX&SE glassware sets D & D2; part 41333. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.
Catalog No. 50-971-487 Supplier Ace Glass, Inc. Supplier No. 396515
Add to Cart
Add to Cart
Request a Quote
Borosilicate glass replacement triple coil condenser for use in rotary evaporator Model R220EX&SE glassware sets D & D2; part 41333. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.