Learn More
Ace Glass, Inc. Condenser tube, 34/35, hose barb inlet/outlets for 5/16in ID tubing
SDP
Catalog No. 50975892
Encompass_Preferred
34/35 condenser tube with hose barb inlet/outlets for 5/16 I.D. tubing. Used with 2000mL reaction kettle with sublimation apparatus accessories.
Catalog No. 50-975-892 Supplier Ace Glass, Inc. Supplier No. 802707
34/35 condenser tube with hose barb inlet/outlets for 5/16 I.D. tubing. Used with 2000mL reaction kettle with sublimation apparatus accessories.