Learn More
Chemglass Life Sciences 3" Condenser Vent Line Assembly, SILCON MED-X 50A Platinum Cured Silicone Tubing 1/4" ID x 3/8" OD Size 17
SDP
Catalog No. 5000083165
Encompass
Polypropylene housing with stepped barbed connections.Condenser line assembly is individually bagged and ready to use.Includes a50mm 0. 22um PTFE filter with a 1/4-3/8 stepped barb.Manufactured withplatinum cured silicone tubing.For sterile applications the filter can be safelyautoclaved up to 30 times.Each filter is identified with a part number and heatstamped with a lot number.
Catalog No. 50-000-83165 Supplier Chemglass Life Sciences Supplier No. CLS139012
Polypropylene housing with stepped barbed connections.Condenser line assembly is individually bagged and ready to use.Includes a50mm 0. 22um PTFE filter with a 1/4-3/8 stepped barb.Manufactured withplatinum cured silicone tubing.For sterile applications the filter can be safelyautoclaved up to 30 times.Each filter is identified with a part number and heatstamped with a lot number.