Learn More
Chemglass Life Sciences Condenser, West, 270mm, 14/20 Joint, 200mm Jacket Length
SDP
Catalog No. CG122021
Encompass_Preferred
Condenser, West, 270mm, 14/20 Joint, 200mm Jacket Length. Narrow annular space between jacket and inner tube of the West condenser provides for excellent cooling efficiency due to the increased flow rate of water. Condenser has a top standard taper outer joint and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration
Catalog No. CG122021 Supplier Chemglass Life Sciences Supplier No. CG122021
Condenser, West, 270mm, 14/20 Joint, 200mm Jacket Length. Narrow annular space between jacket and inner tube of the West condenser provides for excellent cooling efficiency due to the increased flow rate of water. Condenser has a top standard taper outer joint and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration