Learn More
Ace Glass, Inc. Condenser, West, 300mm jacket, 24/40 st joints top and bottom, Ace-Safe hose connections for 1/4" ID tubing
SDP
Catalog No. 50972734
Encompass_Preferred
Condenser, West, 300mm jacket, 24/40 st joints top and bottom, Ace-Safe hose connections for 1/4" ID tubing
Catalog No. 50-972-734 Supplier Ace Glass, Inc. Supplier No. 6029115
Condenser, West, 300mm jacket, 24/40 st joints top and bottom, Ace-Safe hose connections for 1/4" ID tubing