Learn More
Chemglass Life Sciences Condenser, Dewar Type, 14/20 Joint, 120 x 31mm
SDP
Catalog No. CG1209A22
Encompass_Preferred
Condenser, Dewar Type, 14/20 Joint, 120 x 31mm. Condenser has a large opening at the top for use with dry ice/acetone or liquid nitrogen. Condenser has an outer standard taper joint set at a 75º angle to the condenser body and a lower inner drip tip joint, both being the same size.
Catalog No. CG1209A22 Supplier Chemglass Life Sciences Supplier No. CG1209A22
Condenser, Dewar Type, 14/20 Joint, 120 x 31mm. Condenser has a large opening at the top for use with dry ice/acetone or liquid nitrogen. Condenser has an outer standard taper joint set at a 75º angle to the condenser body and a lower inner drip tip joint, both being the same size.