Learn More
Grainger Superior Abrasives Condition Belt, A/O, Coarse, 2.5 in. x 60 in., 4/Pk.
Catalog No. 502758082
Encompass_Preferred
These surface conditioning belts are designed for effective finishing and decorating of large surface areas. The open construction resists loading, overheating, and part discoloration, ensuring consistent results during operation. Engineered with additional flex, these belts easily conform to small contact wheels and narrow workpiece areas.
  • Resists overheating, loading, and discoloration for consistent quality
  • Flexible construction allows for easy conformity to small contact wheels and narrow areas
  • Low stretch reinforcement provides stability and reduces changeover requirements on large or robotic equipment
  • Reduced heat generation promotes long service life
  • Features coarse grade aluminum oxide abrasive for effective material processing
Catalog No. 50-275-8082 Supplier Grainger Supplier No. 152N55
These surface conditioning belts are designed for effective finishing and decorating of large surface areas. The open construction resists loading, overheating, and part discoloration, ensuring consistent results during operation. Engineered with additional flex, these belts easily conform to small contact wheels and narrow workpiece areas.
  • Resists overheating, loading, and discoloration for consistent quality
  • Flexible construction allows for easy conformity to small contact wheels and narrow areas
  • Low stretch reinforcement provides stability and reduces changeover requirements on large or robotic equipment
  • Reduced heat generation promotes long service life
  • Features coarse grade aluminum oxide abrasive for effective material processing