Learn More
Stoelting™ Conditioned Place Preference
SDP
Catalog No. 10000362 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Catalog No. Animal
10-000-362 Mouse
1 options
Catalog No. 10-000-362 Supplier Stoelting™ Supplier No. 64101
Add to Cart
Add to Cart
Stoelting™ Conditioned Place Preference is most commonly used in the assessment of the rewarding property of drugs (or other natural reinforcers) or to identify the neuronal basis of reinforcement expression.
Typically, a drug is injected, and the animal is placed in a test chamber with distinctive environmental cues. This procedure is repeated for several days.
Specifications
For Use With (Application) Neuroscience
Animal Mouse