Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conditioning Reagent, For Sulfate, LabChem™

Catalog No. LC133201
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC133201 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC133201 Supplier LabChem, Inc. Supplier No. LC133201
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Conditioning Reagent
Color Colorless
Name Note For Sulfate
CAS Max % 50.66, 20, 15, 10, 4.34
CAS 67-63-0, 7647-14-5, 56-81-5, 7647-01-0
Linear Formula NaCl
UN Number UN1789
Solubility Information Soluble in water
Formula Weight 58.44
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
Quantity 500 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.