Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Condom Training Model
SDPSCI_ED

Catalog No. S00286 Shop All 3B Scientific Products
Change view
Click to view available options
Color:
Dark Skin Tone
White Skin Tone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S00286 Dark Skin Tone
S00609 White Skin Tone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S00286 Supplier 3B Scientific™ Supplier No. 1000341
Add to Cart
Add to Cart
This item is not returnable. View return policy

3B Scientific Condom training Model of an erect penis with testicles is a great tool for sex education!

  • This model of an erect penis with testicles is used to learn how to use a condom safely.
  • The anatomical structures and its firmness are absolutely realistic, so that your students can train putting on and removing a condom in a realistic way
  • Supplied with 12 dry training condoms and a carrying bag.
  • This is a great tool for sex education.

Specifications

Product Type Condom Training Model
For Use With (Application) Sex Education
Color Dark Skin Tone
Dimensions (L x W x H) 3.0 x 3.0 x 7.7 in.
Gender Male
Includes 12 Dry Training Condoms and a Carrying Bag
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.