Learn More
Grainger Safety Glasses, Uncoated
Catalog No. NC1172977
Encompass_Preferred
Uncoated safety glasses that provide impact protection. They feature polycarbonate lenses that are lightweight and distortion-free. The glasses have adjustable temples and a soft rubber nosepiece for a comfortable fit. They meet ANSI Z87.1 standards for high impact protection. The lenses filter 99.9% of UV rays. These safety glasses are ideal for construction, manufacturing, and other work environments where eye protection is needed.
This item is not returnable. View return policy
Catalog No. NC1172977 Supplier Grainger Supplier No. 4EY97
Uncoated safety glasses that provide impact protection. They feature polycarbonate lenses that are lightweight and distortion-free. The glasses have adjustable temples and a soft rubber nosepiece for a comfortable fit. They meet ANSI Z87.1 standards for high impact protection. The lenses filter 99.9% of UV rays. These safety glasses are ideal for construction, manufacturing, and other work environments where eye protection is needed.