Learn More
Cole-Parmer Oakton Conductivity and TDS Standard, 23 µS; 500 mL
Supplier:  Cole-Parmer 0065323
Encompass_Preferred
• KCl, NaCl and 442 conversions printed on the bottle for ease of use (excluded on 60-mL bottle, but included in insert) • Temperature corrections are printed on the bottle, with certified values listed in tables and referenced to 25°C (excluded on 60-mL bottle, but included in insert) • Manufactured under ISO 9000 quality guidelines and traceable to NIST standards • If unopened, shelf life is 1 year. If opened recommended shelf life is 4 months • Conductivity options range from 1 µS/cm to 200,000 µS/cm
Catalog No. NC9971869