Learn More
Hach Company Conduct Alg Sens-Con 0.5-SS-T
Catalog No. 5000208001 Shop All Hach Company Products
Encompass
Conductivity Sensor-Cell Constant 0 5-Stainless Steel-T-20 ft CableNominal Cell Constant (K) 0 5 Extended Sensor Body with 316 Stainless Steel Outer Electrode Requires 316 SS Ball Valve Assembly MH114M3C 6m (20 ft) Analog Cable Titanium Electrode
Catalog No. 50-002-08001 Supplier Hach Company Supplier No. 3422B6A
Conductivity Sensor-Cell Constant 0 5-Stainless Steel-T-20 ft CableNominal Cell Constant (K) 0 5 Extended Sensor Body with 316 Stainless Steel Outer Electrode Requires 316 SS Ball Valve Assembly MH114M3C 6m (20 ft) Analog Cable Titanium Electrode