Learn More
Hach Company Conduct Sens-Cons 0.5-SS HD
Catalog No. 5000207731 Shop All Hach Company Products
Encompass
Conductivity Sensor-Cell Constant 0 5-Stainless Steel HDNominal Cell Constant (K) 0 5 3/4 inch male NPT threaded 316 stainless steel body 316 Stainless Steel Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode
Catalog No. 50-002-07731 Supplier Hach Company Supplier No. 3444B8D
Conductivity Sensor-Cell Constant 0 5-Stainless Steel HDNominal Cell Constant (K) 0 5 3/4 inch male NPT threaded 316 stainless steel body 316 Stainless Steel Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode