Learn More
Hach Company Conduct Sens-Cons 5.0-SS-T
Catalog No. 5000207742 Shop All Hach Company Products
Encompass
Conductivity Sensor-Cell Constant 5 0-Stainless Steel-T-20 ft CableNominal Cell Constant (K) 5 0 Extended Sensor Body with 316 Stainless Steel Outer Electrode (requires 316 SS Ball Valve Assembly MH114M3C) 6m (20 ft) Analog Cable Titanium Electrode
Catalog No. 50-002-07742 Supplier Hach Company Supplier No. 3422D6A
Conductivity Sensor-Cell Constant 5 0-Stainless Steel-T-20 ft CableNominal Cell Constant (K) 5 0 Extended Sensor Body with 316 Stainless Steel Outer Electrode (requires 316 SS Ball Valve Assembly MH114M3C) 6m (20 ft) Analog Cable Titanium Electrode