Learn More
Hach Company Conduct Sens-PP-Con 0.5-SS-T
Catalog No. 5000207331 Shop All Hach Company Products
Encompass
Conductivity Sensor-Cell Constant 0 5-Stainless Steel-T-PP HDNominal Cell Constant (K) 0 5 Extended Sensor Body with 316 Stainless Steel Outer Electrode (requires 316 SS Ball Valve Assembly MH114M3C) Polypropylene Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode
Catalog No. 50-002-07331 Supplier Hach Company Supplier No. 3422B6B
Conductivity Sensor-Cell Constant 0 5-Stainless Steel-T-PP HDNominal Cell Constant (K) 0 5 Extended Sensor Body with 316 Stainless Steel Outer Electrode (requires 316 SS Ball Valve Assembly MH114M3C) Polypropylene Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode