Learn More
YSI 3P LifeScience CONDUCTIV METER EC300A ECOSENS
Catalog No. NC1475243
Encompass_Preferred
EcoSense EC300A Conductivity MeterInstrument Only. Cables, probes/sensors, and accessories sold separately.Features:1-year instrument, electrode and cable warrantyIP67 waterproof rating; Rugged weighted probe assembly; super-stable electrode designed for field use.Automatic temperature compensationLow battery indicator with 500 hour battery lifeAutomatic shutoff after 30 minutes of inactivity1-, 4- or 10-meter cable lengths availableCE compliance50 data set memory
Catalog No. NC1475243 Supplier YSI 3P LifeScience Supplier No. 606079
EcoSense EC300A Conductivity MeterInstrument Only. Cables, probes/sensors, and accessories sold separately.Features:1-year instrument, electrode and cable warrantyIP67 waterproof rating; Rugged weighted probe assembly; super-stable electrode designed for field use.Automatic temperature compensationLow battery indicator with 500 hour battery lifeAutomatic shutoff after 30 minutes of inactivity1-, 4- or 10-meter cable lengths availableCE compliance50 data set memory