Learn More
Electron Microscopy Sciences Conductive Adhesive Tabs, 2
SDP
Catalog No. 5028177
Encompass_Preferred
An affordable double-sided conductive tab for all SEM samples, these 1/2" (12 mm) diameter conductive tabs work like carbon adhesive tabs. Just remove tab from the roll, press onto surface where you want the film, lift"tab" and peel it off. The tabs can be cut to desired size before being removed from its backing, for use on smaller samples.
They have a smoother background (over 99% transparent to EDS), high strength adhesive, a thickness of only 0.002", and better particle detection. Contains a trace of nickel (<0.6%) and copper (<0.3).
Catalog No. 50-281-77 Supplier Electron Microscopy Sciences Supplier No. 7676201
An affordable double-sided conductive tab for all SEM samples, these 1/2" (12 mm) diameter conductive tabs work like carbon adhesive tabs. Just remove tab from the roll, press onto surface where you want the film, lift"tab" and peel it off. The tabs can be cut to desired size before being removed from its backing, for use on smaller samples.
They have a smoother background (over 99% transparent to EDS), high strength adhesive, a thickness of only 0.002", and better particle detection. Contains a trace of nickel (<0.6%) and copper (<0.3).