Learn More
Electron Microscopy Sciences Scotch™ Conductive Aluminum Tape, 1/4" wide 18 YD
SDP
Supplier:  Electron Microscopy Sciences 77800
Encompass_Preferred
The conductive tape is used in scanning electron microscopy specimen mounting. Aluminum foil tape (Scotch 1170) with conductive adhesive. 3" core and foil thickness: 2 mils. Total thickness: 4mils (.102mm), adhesion: 35oz/in, electrical resistance through adhesive: .01ohm/in(.07ohm/cm)
Width: 1/4"
Catalog No. 50-285-21