Learn More
Electron Microscopy Sciences GEL-TRAY Conductive BD Tray in Transparent Hinged Box, 2" x 2"
SDP
Catalog No. 501900240
Encompass
Intended for manual applications in which the devices are loaded and unloaded using tweezers or by fingers. Configured as a 2" carrier stored in a protective plastic hinged box. The Gel coating is applied directly to the surface of the plastic tray. Can be easily removed from the box allowing for convenient handling when loading or unloading fixtures are being used. A removable 2" Gel coated plastic tray coated inside a hinged plastic box.
Type: Transparent Hinged
Size: 2" x 2"
Catalog No. 50-190-0240 Supplier Electron Microscopy Sciences Supplier No. 6490014
Intended for manual applications in which the devices are loaded and unloaded using tweezers or by fingers. Configured as a 2" carrier stored in a protective plastic hinged box. The Gel coating is applied directly to the surface of the plastic tray. Can be easily removed from the box allowing for convenient handling when loading or unloading fixtures are being used. A removable 2" Gel coated plastic tray coated inside a hinged plastic box.
Type: Transparent Hinged
Size: 2" x 2"