Learn More
Electron Microscopy Sciences EMS Conductive Carbon Paint #502 30 GR
SDP
Catalog No. 50365692
Encompass_Preferred
Conductive adhesive 502 is a high temperature paint that is a combination of specially processed carbon particles in a fluoroelastomer resin system designed to provide high resistance values. In its cured form, it exhibits both high and low temperature flexibility and moisture resistance.
Features
Withstands ambient temperatures of over 500 degree F (260 degree C).
Remains flexible over temperature range of –40 degree F to over 500 degree F (260 degree C).
Cures at room temperature.
Good adhesion to a variety of substrate.
Excellent oxidation resistance.
Ready to use and easy to apply
Catalog No. 50-365-692 Supplier Electron Microscopy Sciences Supplier No. 1269430
Conductive adhesive 502 is a high temperature paint that is a combination of specially processed carbon particles in a fluoroelastomer resin system designed to provide high resistance values. In its cured form, it exhibits both high and low temperature flexibility and moisture resistance.
Features
Withstands ambient temperatures of over 500 degree F (260 degree C).
Remains flexible over temperature range of –40 degree F to over 500 degree F (260 degree C).
Cures at room temperature.
Good adhesion to a variety of substrate.
Excellent oxidation resistance.
Ready to use and easy to apply