Learn More
Electron Microscopy Sciences Scotch™ Conductive Copper Tape, 1/4" wide 18 YD
SDP
Catalog No. 5028519
Encompass_Preferred
The conductive tape is used in scanning electron microscopy specimen mounting. Copper foil tape (Scotch 1181), with conductive adhesive, 3" core and foil thickness: 1.4 mils. Total thickness: 3mils (.076mm), adhesion: 35oz/ln (3.81 N/10mm), electrical resistance: .005ohm/in(.032ohm/cm)
Width: 1/4"
Catalog No. 50-285-19 Supplier Electron Microscopy Sciences Supplier No. 77802
The conductive tape is used in scanning electron microscopy specimen mounting. Copper foil tape (Scotch 1181), with conductive adhesive, 3" core and foil thickness: 1.4 mils. Total thickness: 3mils (.076mm), adhesion: 35oz/ln (3.81 N/10mm), electrical resistance: .005ohm/in(.032ohm/cm)
Width: 1/4"