Learn More
Electron Microscopy Sciences Conductive Silver Pen
SDP
Catalog No. 50980316
Encompass_Preferred
This pen is designed for making instant conductive silver traces. It is ideal for applying samples to SEM stubs. A unique valve tip allows for very smooth flow withnormal writing pressure and it is spring loaded to prevent clogging. For conductivity traces, solderable termination's are possible using a 250 degree F cure for 15-20 minutes. Tin, lead, or silver solder can be used (Do not exceed 350 degree F for more than 5 seconds).
Each pen is filled with approximately 100 feet of traces. Silver content: 39-45% with less than 10 micron diameter. The thinner used for this pen is Butyl Acetate.
Catalog No. 50-980-316 Supplier Electron Microscopy Sciences Supplier No. 1264401
This pen is designed for making instant conductive silver traces. It is ideal for applying samples to SEM stubs. A unique valve tip allows for very smooth flow withnormal writing pressure and it is spring loaded to prevent clogging. For conductivity traces, solderable termination's are possible using a 250 degree F cure for 15-20 minutes. Tin, lead, or silver solder can be used (Do not exceed 350 degree F for more than 5 seconds).
Each pen is filled with approximately 100 feet of traces. Silver content: 39-45% with less than 10 micron diameter. The thinner used for this pen is Butyl Acetate.