Learn More
Sigma Aldrich Conductive silver printing ink, resistivity 9 - 10 muOmega-cm
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Specifications
Specifications
| Quantity | 10 g |
| Synonym | Dyesol™ DYAG100 conductive silver ink; Screen printable silver paste |
| Percent Purity | 75-85% (Solids content) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.