Learn More
Aqua Solutions CONDUCTIVIT STD 1413UMHOS 500M
SDP
Catalog No. NC0441195
Encompass_Preferred
CONDUCTIVITY STANDARD 0.01 MOLAR (1,413 uMHOS @25 deg.C) N.I.S.T TRACEABLE (500ML)
Catalog No. NC0441195 Supplier Aqua Solutions Supplier No. 6920500ML
CONDUCTIVITY STANDARD 0.01 MOLAR (1,413 uMHOS @25 deg.C) N.I.S.T TRACEABLE (500ML)