Learn More
Aqua Solutions CONDUCTIVITY 1000 UMHOS 1 L
SDP
Catalog No. NC9593189
Encompass_Preferred
Conductivity Standard Traceable to N.I.S.T., 100 micrometerho, 1 L, BP: 100degreeC, Water Soluble, Appearance and Odors: Clear water white/no odor, Specific Gravity: 1, Volatile by Volume: >99%
Catalog No. NC9593189 Supplier Aqua Solutions Supplier No. 69501L
Conductivity Standard Traceable to N.I.S.T., 100 micrometerho, 1 L, BP: 100degreeC, Water Soluble, Appearance and Odors: Clear water white/no odor, Specific Gravity: 1, Volatile by Volume: >99%