Learn More
YSI 3P LifeScience Conductivity Calibrator, 50,000 umhos/cm, 8/pk
Catalog No. 50200110
Encompass_Preferred
NIST-traceable conductivity solutions to calibrate both field instruments and lab instruments and cells together for maximum accuracy.
Catalog No. 50-200-110 Supplier YSI 3P LifeScience Supplier No. 060660
NIST-traceable conductivity solutions to calibrate both field instruments and lab instruments and cells together for maximum accuracy.