Learn More
Cole-Parmer Cole-Parmer Conductivity Cell, K=0.01, 10 Kohm ATC, stainless steel body
Catalog No. 50633456 Shop All Cole-Parmer Products
Encompass_Preferred
These 10-foot cables with stripped ends are economical replacements for a variety of controllers. There are flow-through, in-line, swage, and dip styles available.
Catalog No. 50-633-456 Supplier Cole-Parmer Supplier No. 1950063
These 10-foot cables with stripped ends are economical replacements for a variety of controllers. There are flow-through, in-line, swage, and dip styles available.