Learn More
Cole-Parmer Cole-Parmer C621-3-1-1 Conductivity Cell; K=0.1, SS, In-Line; 100 Ohm Pt RTD ATC
Catalog No. 50633629 Shop All Cole-Parmer Products
Encompass_Preferred
These 10-foot cables with stripped ends are economical replacements for a variety of controllers. There are flow-through, in-line, swage, and dip styles available.
Catalog No. 50-633-629 Supplier Cole-Parmer Supplier No. 1950044
These 10-foot cables with stripped ends are economical replacements for a variety of controllers. There are flow-through, in-line, swage, and dip styles available.