Learn More
Cole-Parmer Cole-Parmer Conductivity Cell, K=10.0, 10 Kohm ATC, PVDF body
Catalog No. 50633459 Shop All Cole-Parmer Products
Encompass_Preferred
These 10-foot cables with stripped ends are economical replacements for a variety of controllers. There are flow-through, in-line, swage, and dip styles available.
Catalog No. 50-633-459 Supplier Cole-Parmer Supplier No. 1950067
These 10-foot cables with stripped ends are economical replacements for a variety of controllers. There are flow-through, in-line, swage, and dip styles available.