Learn More
Cole-Parmer Cole-Parmer Conductivity Cell; K=10; Epoxy, In-line; 100 Ohm Pt RTD ATC
Catalog No. 501961244 Shop All Cole-Parmer Products
Encompass_Preferred
Cells withstand pressures up to 150 psi (at 135C for SS; 25C for CPVC and PVDF).
Catalog No. 50-196-1244 Supplier Cole-Parmer Supplier No. 3582022
Cells withstand pressures up to 150 psi (at 135C for SS; 25C for CPVC and PVDF).