Learn More
Waters Corp WATERS CORPORATION Conductivity Detector OQ/PQ Mobile Phase, for OQ/PQ Procedure, Acetonitrile Formulation
Catalog No. 50792737 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Conductivity Detector OQ/PQ Mobile Phase is designed for use with the Conductivity Detector to detect changes in the thermal conductivity of column eluent compared to a reference flow. This lab equipment is specifically the Mobile Phase utilized during the OQ/PQ procedure. It contains acetonitrile, which is a main component of the solution.
  • Specifically formulated for the conductivity OQ/PQ procedure.
  • Aids in detecting changes in thermal conductivity between column eluent and a reference flow.
  • Contains acetonitrile as one of its main solution components.
Catalog No. 50-792-737 Supplier Waters Corp Supplier No. 700002310
The Conductivity Detector OQ/PQ Mobile Phase is designed for use with the Conductivity Detector to detect changes in the thermal conductivity of column eluent compared to a reference flow. This lab equipment is specifically the Mobile Phase utilized during the OQ/PQ procedure. It contains acetonitrile, which is a main component of the solution.
  • Specifically formulated for the conductivity OQ/PQ procedure.
  • Aids in detecting changes in thermal conductivity between column eluent and a reference flow.
  • Contains acetonitrile as one of its main solution components.