Learn More
Squishy Circuits™ Conductivity Kit
Description
- Deluxe Kit - The most complete kit that includes a deluxe battery holder, which has a knob that allows your lights to blink and buzzers to beep at different speeds
- Contents: 1 x Deluxe Battery Holder, 1 x Piezoelectric Buzzer, 1 x Motor with Fan, 1 x Mechanical Buzzer, 1 x Switch, 40 x LEDs (5 each or Red, Green, White, Yellow, Blue, Purple, Orange, and Color Changing), 1 x Set of Dough Sculpting Tools, 1 x Rolling Pin, 1 x 7 oz. Container of White Insulating Dough, 7 x 7 oz. Containers of Conductive Dough (in Red, Blue, and Green, Yellow, Orange, Purple, and Black)
- Dough Kit - Designed to make play dough science projects fun through an array of different colors
- Contents: 7 containers of conductive dough and 1 container of white insulating dough
- Each container holds 7 ounces
- The dough is safe, non-toxic, and does contain gluten
- The conductive dough dries out if left out of the container
- The insulating dough does not dry out
- Group Kit - Ideal for classroom groups of up to 30 peoples
- Contents: 10 x Battery Holders (40 AA Batteries not Included), 10 x Piezoelectric Buzzers, 10 x Motors with Fan Blades, 10 x Switches, 80 x LEDs (10 each in Red, Green, White, Yellow, Orange, Purple, Blue, and Color Changing)
- Kit does not come with dough
- Lite Kit - Basic kit that is ideal for classrooms and events where every participant needs their own battery pack
- Contents: 1 x Battery Holder, 15 x LEDs (5 each or Red, Green, Blue), 1 x 7 oz. Container of Conductive Dough (colors vary)
- Standard Kit - Includes everything needed to get started with more advanced projects
- Contents: 1 x Battery Holder, 1 x Piezoelectric Buzzer, 25 x LEDs (5 each or Red, Green, White, Yellow, Blue), 1 x 3.5 oz. Container of Insulating Dough, 3 x 3.5 oz. Containers of Conductive Dough (in Red, Green, and Blue)
Specifications
Specifications
| Age | 8 |
| Quantity | 1 Kit |
| Product Type | Deluxe Kit |