Learn More
Hach Company Intellical™ CDC401 Laboratory 4-Poles Graphite Conductivity Cell, 3 m Cable
Catalog No. 501997555 Shop All Hach Company Products
Encompass
Intellical™ CDC401 is a digital, graphite, 4-pole conductivity cell with a temperature sensor. The standard version is available with a 3 or 1 m cable and is intended for laboratory use. The CDC401 is ideal for measuring electrical conductivity, salinity, resistivity, or total dissolved solids (TDS) in wastewater, drinking water, or general water quality applications.
Catalog No. 50-199-7555 Supplier Hach Company Supplier No. CDC40103
Intellical™ CDC401 is a digital, graphite, 4-pole conductivity cell with a temperature sensor. The standard version is available with a 3 or 1 m cable and is intended for laboratory use. The CDC401 is ideal for measuring electrical conductivity, salinity, resistivity, or total dissolved solids (TDS) in wastewater, drinking water, or general water quality applications.