Learn More
Hach Company Contacting Conductivity Sensor for low conductivity (k=0.05) with 1/2" Kynar Compression Fitting
Catalog No. 501997437 Shop All Hach Company Products
Encompass
Nominal Cell Constant (K) = 0.05, 1/2 inch NPT Kynar (PVDF) Compression Fitting, 6m (20ft) Analog Cable, Titanium Electrode
Catalog No. 50-199-7437 Supplier Hach Company Supplier No. 3422A1A
Nominal Cell Constant (K) = 0.05, 1/2 inch NPT Kynar (PVDF) Compression Fitting, 6m (20ft) Analog Cable, Titanium Electrode