Learn More
Hach Company Contacting Conductivity Sensor, High Conductivity, K=5.0, 3/4" Kynar Compression Fitting
Catalog No. 501997440 Shop All Hach Company Products
Encompass
Nominal Cell Constant (K) = 5.0, 3/4 inch NPT Kynar (PVDF) Compression Fitting, 6m (20ft) Analog Cable, Titanium Electrode
Catalog No. 50-199-7440 Supplier Hach Company Supplier No. 3422D3A
Nominal Cell Constant (K) = 5.0, 3/4 inch NPT Kynar (PVDF) Compression Fitting, 6m (20ft) Analog Cable, Titanium Electrode