Learn More
Taylor Chemical CONDUCTIVITY SOLTN 1000US,16OZ
Catalog No. NC2130776
Encompass_Preferred
Conductivity Solution 1000 μS (492 ppm NaCl), 16 oz
This item is not returnable. View return policy
Catalog No. NC2130776 Supplier Taylor Chemical Supplier No. R08681KE
Conductivity Solution 1000 μS (492 ppm NaCl), 16 oz