Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack
Specifications
Specifications
| Chemical Name or Material | Conductivity Standard |
| Packaging | Natural Poly Bottle |
| Name Note | 1.413mS/cm (1413 μmho/cm) at 25°C |
| CAS | 7647-14-5, 7732-18-5 |
| CAS Min % | 98.0, 0.07 |
| CAS Max % | 100.0, 0.07 |
| Odor | Odorless |
| Physical Form | Liquid |
| Color | Colorless |
| pH | 6.7 to 7.3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction