Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 1000 μS/cm at 25°C (495 ppm TDS as NaCl), Ricca Chemical
SDP

Catalog No. 2243100P Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
20 x 20 mL
4 L
Packaging:
Natural Poly Bottle
Pack
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2243100P 100 mL Pack
224320 20 x 20 mL Pack
2243-16 500 mL Natural Poly Bottle
2243-32 1 L Natural Poly Bottle
2243-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 2243100P Supplier Ricca Chemical Company Supplier No. 2243100P
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Pack
Name Note 1mS/cm (1000μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 98.0, 0.05
CAS Max % 100.0, 0.05
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 495ppm as NaCl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.