Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 100,000 μS/cm (100 mS/cm) at 25°C, Ricca Chemical
SDP

Catalog No. 22491001 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22491001 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22491001 Supplier Ricca Chemical Company Supplier No. 2249.1001
Add to Cart
Add to Cart

65,750 ppm TDS as NaCl

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
CAS 7647-14-5, 7732-18-5
CAS Min % 92.0, 6.31
CAS Max % 95.0, 6.56
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Melting Point Approximately 0°C
Boiling Point Approximately 100.1°C
For Use With (Equipment) Conductivity Meters and Probes
Solubility Miscible with water
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 4 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 100mS/cm (100, 000μmho/cm) at 25°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.