Learn More
Aqua Solutions CONDUCTIVITY STANDARD 100UMHOS
SDP
Catalog No. NC9380369
Encompass_Preferred
Conductivity Standard Traceable to N.I.S.T.; Clear water white; 500mL; M.P. ~0 deg.C; B.P. ~100 deg.C; Specific Gravity: 1
Catalog No. NC9380369 Supplier Aqua Solutions Supplier No. 6919500ML
Conductivity Standard Traceable to N.I.S.T.; Clear water white; 500mL; M.P. ~0 deg.C; B.P. ~100 deg.C; Specific Gravity: 1