Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 130,000 μS/cm (130 mS/cm) at 25°C, Ricca Chemical
SDP

Catalog No. 22491316 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22491316 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22491316 Supplier Ricca Chemical Company Supplier No. 2249.1316
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
CAS 7647-14-5, 7732-18-5
CAS Min % 90.0, 7.7
CAS Max % 94.0, 8.02
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Melting Point Approximately 0°C
Boiling Point Approximately 100.1°C
For Use With (Equipment) Conductivity Meters and Probes
Solubility Miscible with water
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.