Learn More
Cole-Parmer Oakton Traceable Conductivity and TDS Standard, Batch-Tested, 10,000 µS; 500 mL
Catalog No. NC9556866 Shop All Cole-Parmer Products
Encompass_Preferred
All Oakton Traceable conductivity standards are NIST-traceable reference materials, supplied with a calibration report from an ISO 17034 and ISO 17025 accredited laboratory. These standard solutions let you calibrate conductivity instruments to ensure accuracy. If unopened, shelf life is one year; once opened, shelf life will vary by solution value. Use when readings are expected to be around 10,000 S/cm.
Catalog No. NC9556866 Supplier Cole-Parmer Supplier No. 0065232
All Oakton Traceable conductivity standards are NIST-traceable reference materials, supplied with a calibration report from an ISO 17034 and ISO 17025 accredited laboratory. These standard solutions let you calibrate conductivity instruments to ensure accuracy. If unopened, shelf life is one year; once opened, shelf life will vary by solution value. Use when readings are expected to be around 10,000 S/cm.