Learn More
Conductivity Standard - 2,060 μmhos/cm, MilliporeSigma™ Supelco™
Catalog No. 111004896 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
11-100-4896 500 mL
1 options
Catalog No. 11-100-4896 Supplier MilliporeSigma Supelco Supplier No. COND2060500ML
Add to Cart
Add to Cart
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Featured Industries: Environmental
TRUSTED_SUSTAINABILITY
Specifications
Type Certified Reference Material
Quantity 500 mL
Grade Certified Reference Material