Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 20,000 μS/cm (20 mS/cm) at 25°C (11,000 ppm TDS as NaCl), Ricca Chemical
SDP

Catalog No. 224732 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2247-32 1 L
2247-16 500 mL
2247-1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2247-32 Supplier Ricca Chemical Company Supplier No. 224732
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Natural Poly Bottle
CAS 7647-14-5, 7732-18-5
Concentration or Composition Notes 11,000ppm as NaCl
Physical Form Liquid
Molecular Formula ClNa
MDL Number MFCD00003477
Color Colorless
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
Quantity 1 L
PubChem CID 5234
ChEBI CHEBI:26710
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.